Digital Sensor Extension Cable
Supplier: Vernier Software & Technology
EXT-BTD
470219-278EA 12.59 USD
470219-278
Digital Sensor Extension Cable
Cables
The Digital Sensor Extension Cable can be used with any Vernier sensor that connects to a digital channel of one of our interfaces. The port will be labeled DIG1 or DIG2.