Quartz Cuvettes
Supplier: Vernier Software & Technology
CUV-QUARTZ
470219-264PK 321.89 USD
470219-264
Quartz Cuvettes
Cuvettes Spectrophotometer Cuvettes
For use with UV-VIS Spectrophotometers
  • Standard rectangluar cuvettes
  • 170–2700nm spectral range
  • Two polished sides
The Quartz Cuvettes are a pack of two Spectrosil® Quartz cuvettes for use with UV-VIS spectrophotometers. They are the same pack of cuvettes that ship with the Vernier UV-VIS Spectrophotometer.