Vernier® Replacement Blood Pressure Cuffs
Supplier: Vernier Software & Technology
CUFF-LG CUFF-SM CUFF-STD
470219-256EA 41.99 USD
470219-256 470219-258 470219-260
Vernier® Replacement Blood Pressure Cuffs
First Aid Supplies Surgical Drapes
Easily measure blood pressure in the classroom.
  • Three sizes available
  • No stethoscope needed
  • Cuff interfaces directly with Vernier® blood pressure sensors
  • Not compatible with standard sphygmomanometers
These blood pressure cuffs are replacements for use only with the Vernier® blood pressure sensors. Compatible with either the BTA or the Go Direct Vernier® sensors.
Ordering information: Required but not included: Vernier® Blood pressure sensor.