Formaldehyde Solution Spill Kit
Supplier: Ward's Science
IS5035
470219-190EA 68.05 USD
470219-190
Formaldehyde Solution Spill Kit
Spill Control Materials
Be prepared for spills in your lab.
  • Enough Material to Clean One Liter of a 10% Formalin Solution
  • Instructions and SDS
  • Vinyl Exam Gloves
This kit includes a dust pan and brush to properly dispose of spills.
It also includes vinyl exam gloves, so you are protected while cleaning.
Ordering information: This kit includes an instruction sheet/SDS, vinyl exam gloves, polybags, twist ties, blank shipping tags, dust pan and brush, vermiculite, and an absorbent formaldehyde neutralizing agent (sodium bisulfite).