Narrow Mouth Reagent Bottles, with Screw Cap
Supplier: DWK Life Sciences (KIMBLE)
220283
470219-160CS 142.35 USD
470219-160
Narrow Mouth Reagent Bottles, with Screw Cap
Bottles Roller Bottles
These Reagent bottles are made of borosilicate glass with polopropylene screw caps with PTFE liner. They feature a no-drip pour lip and a written label area. Autoclavable.