Sterile Centrifuge Tubes, Tarsons
Supplier: VWR International
470219-158
470219-158CS 247.85 USD
470219-158
Sterile Centrifuge Tubes, Tarsons
Tubes Centrifuge Tubes
Conical bottom, polypropylene centrifuge tubes are supplied with cap.
Tubes are sterile and autoclavable. Length: 120 mm. Capacity: 15 ml.