Stainless Steel Lab Scoops, Eisco Scientific
Supplier: Wards
CH0635A
470219-130EA 3.5 USD
470219-130
Stainless Steel Lab Scoops, Eisco Scientific
Scoops
Durable and versatile lab scoops for classroom or laboratory.
  • Lab scoops/spatulas
  • 6.5" long
Many uses in science laboratories and classrooms. These stainless steel scoops features one pointed end, one rounded end, and semicircular cross section.