Element Groups
Supplier: Ward's Science
IS2516
470219-084EA 179.75 USD
470219-084
Element Groups
Educational Materials Chemistry Educational Materials Periodic Table Learning Activities
The activities in this kit focus on developing a better understanding of why periodic trends occur in the elements.
  • Kit teaches students to look for patterns as they conduct experiments
  • Students learn why the periodic table looks as it does
  • Graduated plastic pipette included
  • Two sets of experiments
Students perform two sets of experiments focusing on the alkaline earth metals and the halogens groups. In the first set of experiments, students search for patterns as they look at the reactivity of the alkaline earth metals.
The second set of experiments form small amounts of solubilized halogen gasses as a colorful way of measuring reactivity within the halogen group.
Ordering information: This kit includes ammonium oxalate, barium nitrate, calcium nitrate, chlorine water, iodine water, magnesium nitrate, mineral oil, potassium bromate, potassium bromide, potassium bromide, potassium iodate, sodium carbonate, potassium chloride, potassium iodide, strontium nitrate, sulfuric acid, sulfuric acid, graduated plastic pipette, and vials and labels for bromine water.
Delivery information: Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.