CENCO® Second Law of Motion Apparatus
Supplier: Wards
0877R
470110-836EA 115 USD
470110-836 470218-760
CENCO® Second Law of Motion Apparatus
Educational Materials Physics Educational Materials Mechanics Learning Activities
This apparatus provides an exciting example of Newton's Second Law.
  • Heavy Cast Metal Base Prevents Unnecessary Movement
  • Two Spring Setting Vary the Launch Velocity
  • For Demonstrating Newton's Second Law
  • Two Steel Ball Provided
  • Instructions Provided
The unit is based on a substantial metal casting, and the entire mechanism is visible. The spring gun offers two tensions for two launch velocities.
The 25 cm long unit has a 10 mm rod.
Ordering information: This kit includes two steel balls and instructions. A support stand and clamp are required but not included.