FDA Approved Autoclave with Printer
Supplier: Revolutionary Science
RS-SC-200P
470218-628EA 2573.15 USD
470218-628
FDA Approved Autoclave with Printer
Sterilizers Autoclaves (Steam Sterilizers)
Simple push-button operation
Digital display
Stainless steel chamber and rugged ABS enclosure
Front loading door
Will not corrode like aluminum
Dry mode
Two-year warranty
Lightweight and portable
Robust
Built in America out of stainless steel and rugged ABS
Simple one-button automatic operation
ST79 approved sterilizing cycle
Dry mode allows users to sterilize dry wrapped instruments
Temperature, time, and pressure on digital display
Stainless steel chamber will not corrode like aluminum
Printer included with device
Under and over temperature alarms
Insufficient water alarm
Silicone rubber gasket, assures a long life