FDA Approved Front Loading Autoclave
Supplier: Revolutionary Science
RS-SC-200
470218-626EA 2263.5 USD
470218-626
FDA Approved Front Loading Autoclave
Sterilizers Autoclaves (Steam Sterilizers)
This front-loading autoclave features simple push-button operation and a digital display.
  • Dry Mode Allows Users to Sterilize Dry-Wrapped Instruments
  • Temperature, Time, and Pressure on Digital Display
  • Stainless Steel Chamber Doesn't Corrode Like Aluminum Does
  • Printer Access Port for Optional Printer or Computer Connection
  • Under and Over-Temperature Alarms
The stainless steel chamber and rugged ABS enclosure don't corrode the way aluminum can, and the silicone rubber gasket ensures a long life.
The dry mode allows users to sterilize dry-wrapped instruments. The printer-access port lets users connect the autoclave to printers or computers.
Certifications: ST79-approved sterilizing cycle.
Ordering information: This kit includes a two-year manufacturer's warranty.