FDA Approved Top Loading Autoclave, 8L
Supplier: Revolutionary Science
RS-SC-102
470218-624EA 1556.15 USD
470218-624
FDA Approved Top Loading Autoclave, 8L
Sterilizers Autoclaves (Steam Sterilizers)
The FDA-approved, Saniclave 102 is fashioned with a simple push-button operation.
  • 8-liter stainless steel chamber
  • Sterilizes solid instruments, great for microbiology, cooking media or agarose, garbage, general laboratory and veterinary use
  • Standard 121 ºC, 30 minutes
  • Lightweight, portable, safe burn-free exterior
  • Approved for human medical use
The thermally protective polymer skin eliminates burn risk. Low water and under and over-temperature alarms assure safety and effectiveness during operation. A corrosion-resistant, stainless steel chamber makesMore Product Information