Accessories for Ward's Science Vehicle Accessory Kit
Supplier: Avantor
470218-424KT 198.59 USD
470218-424
Accessories for Ward's Science Vehicle Accessory Kit
Educational Materials Physics Educational Materials
The accessory kit is available to explore solar and electrical power as alternative sources and includes:
8 small worm gears for 2mm shaft
8 gears with bushing
8 electric motors, 1.5-3V
8 motor pulleys
8 self adhesive clips
1 9V solar cell, 150mA
8 9V batteries
8 9V battery holders, steel
16 pieces insulated wire, 22 gauge, with stripped ends
8 slide switches