Photosynthesis, The Hill Reaction
Supplier: Ward's Science
IS3500
470218-356EA 98.8 USD
470218-356
Photosynthesis, The Hill Reaction
Educational Materials Biology Educational Materials Plant Learning Activities
Learn About The Light Cycle In Photosynthesis
  • Perform Experiments Based On The Hill Reaction
  • Includes All Necessary Chemicals
  • Includes Teachers Manual And Student Guide
The ability to convert light energy into usable chemical energy (photosynthesis) makes life possible. The process of photosynthesis can be broken down into two parts known as light and dark cycles. The activities in this kit focus on understanding the function of the light cycle, by running experiments based on the Hill reaction. The instructions are written for both visual analysis and/or UV-Vis analysis. The kit contains material for six groups.
Included:
propylene glycol solution
plant extract buffer
dichloroindophenol EZ prep
sucrose EZ prep
sodium hydrosulfite
filters
graduated pipettes
Teachers Manual
Student Copy Master