Dry Bath Digital With Isolated Chambers
Supplier: Ward's
BSH6000
470216-346EA 1088.5 USD
470216-346
Dry Bath Digital With Isolated Chambers
Block Heaters Digital Block Heaters
Digital dry baths that include timed or continuous operations and a removable hinged lid. External temperature probe can be positioned directly inside a sample tube.
  • Two isolated chambers (independent control of time & temp.)
  • Digital temperature control (set and walk away)
  • Hinged lid improves uniformity & conserves energy
  • Blocks sold separately