Electric Bell
Supplier: VWR International
PH1030B
470213-396EA 18.1 USD
470213-396
Electric Bell
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Electric bell that operated on 4 to 6 Volts DC with detachable cover and metal thread terminals.
Bell is approximately 75 in diameter.