Eisco® Enlarged Human Lower Jaw with Teeth, 3 Parts
Supplier: Wards
AM234AS
470212-456EA 175.39 USD
470212-456
Eisco® Enlarged Human Lower Jaw with Teeth, 3 Parts
Models Human Biology Models
An enlarged and highly detailed model of the left side of a human lower jaw, demonstrating external and internal anatomical features.
  • Enlarged (5x) model includes left side lower jaw with adult teeth
  • 3 piece model can be taken apart
  • Hand painted
  • Rod mounted on base
  • Includes numbered key card
This highly detailed model of the left side of a human lower jaw is enlarged 5 times life size. It includes all of the adult teeth and a cut-away of the body of the mandible to show nerves and blood vessels going to the teeth. An additional incisor tooth is bisected to show internal structures.