Self-Zeroing Automatic Buret Kit
Supplier: Thermo Fisher Scientific
3645-2550
470211-802CS 267.3 USD
470211-802
Self-Zeroing Automatic Buret Kit
Burets
Nalgene® This kit makes any 25 or 50 mL buret both auto-filling and self-zeroing. With 1000 mL squeeze bottle, 1.2 m of clear flexible tubing and fill tube with adapter. Buret not included.