PYREX® Ignition Tubes
Supplier: Corning
9860-16
470211-684CS 1229.9 USD
470211-684
PYREX® Ignition Tubes
Tubes Test Tubes
These PYREX® rimless ignition tubes are made from No. 7740 glass.
  • Very good chemical resistance
  • Good temperature resistance
These ignition tubes are satisfactory for most applications encountered in school work where temperatures above 600 °C are not required. These rimless tubes perform well when continually used at temperatures up to 500 °C, or for short periods to 600 °C.