Pyrex® Graduated Centrifuge Tube
Supplier: Corning
8080-15
470211-622CS 251.15 USD
470211-622
Pyrex® Graduated Centrifuge Tube
Tubes Centrifuge Tubes
PYREX® borosilicate glass.
  • Graduated from 0.1 to 15 ml with 0.1 ml interval
Conical bottom with beaded rim.