Ungraduated Pyrex® Centrifuge Tubes
Supplier: Corning
8060-15
470211-616CS 133.55 USD
470211-616
Ungraduated Pyrex® Centrifuge Tubes
Tubes Centrifuge Tubes
Made of PYREX® borosilicate glass, conical, beaded.
  • Wide neck
  • Excellent chemical resistance
  • Excellent temperature resistance
These centrifuge tubes has uniform glass distribution in the wall, taper and bottom provides improved mechanical strength, prevents the formation of failure-causing stress regions during centrifuging and insures the highest degree of accuracy.