PYREX® Measuring Pipettes
Supplier: Corning
7065-2
470211-586CS 248.95 USD
470211-586
PYREX® Measuring Pipettes
Pipettes Measuring (Mohr) Pipettes
Calibrated to deliver, these Mohr-type pipettes are color-coded by size for easy identification.
  • Colored graduations are enameled onto glass
  • Calibrated 'to deliver' (TD) their total capacity without blow-out
Pipette is reusable with round tip.