PYREX® VISTA™ Single Metric Volumetric and Graduated Cylinders, 25 ml, TC, Plastic Hexagonal Base, Blue Graduations
Supplier: Corning
70075-25
470211-580CS 161.35 USD
470211-580
PYREX® VISTA™ Single Metric Volumetric and Graduated Cylinders, 25 ml, TC, Plastic Hexagonal Base, Blue Graduations
Cylinders Measuring Cylinders
Made of PYREX® glass, blue graduations.
  • Calibrated to contain (TC)
  • Tolerances are ±5% of total at any point
  • 10 ml cylinder has a funnel top
They are supplied in two parts - a strong, accurate glass graduate and a detachable, sturdy plastic base which absorbs impacts and reduces breakage. Cylinders can be conveniently stacked when plastic bases are removed.
Delivery information: Supplied with bumper guards for cylinders of capacity 25 to 100 ml. Snap caps are supplied with all sizes.