PYREX® Micro-Organic Chemistry Kit – Deluxe Version
Supplier: Corning
6949M-1
470211-574KT 1078.2 USD
470211-574
PYREX® Micro-Organic Chemistry Kit – Deluxe Version
Microchemistry Kits
This unique glassware kit that is among the finest for use in the microchemistry lab.
  • Feature interchangeable standard taper joints, enabling you to use fewer clamps while performing distillation, extraction reflux and other microscale experiments
All components are made of Pyrex® glass.
Delivery information: Includes sturdy molded polyethylene storage cases with a cut-out foam insert, air reflux condenser (7/10M – 7/10F), air reflux condenser (14/10M – 7/10F), capillary gas delivery tube (7/10M), two gas chromatograph collection tubes (5/5M), Craig recrystallization tube (1 mL capacity), two conical reaction vials (complete, 0.1 mL capacity, 5/5F), conical reaction vial (complete, 0.3 mL capacity, 7/10F) and an additional conical reaction vial (complete, 3.0 mL capacity, 14/10F).