Ward's® 171 Stirrer
Supplier: Corning
6795-171
470211-554EA 256.39 USD
470211-554
Ward's® 171 Stirrer
Stirrers Magnetic Stirrers
A compact, well-priced stirrer for your classroom laboratory.
  • Weighs 1.8 kg
  • Stirs at speeds of 100–1600 rpm
  • One year warranty
  • UL approved
The space-saving design is a plus, and it provides vigorous stirring with an alternating current motor for long life.