PYREX® Separatory Funnels, Squibb
Supplier: Corning
6402-125
470211-548CS 735.6 USD
470211-548
PYREX® Separatory Funnels, Squibb
Funnels Separating Funnels
Designed for Long Life and Maximum Safety
  • Heavy walled, borosilicate glass for maximum mechanical strength
  • HPDE standard taper stopper no.:22
  • Replaceable PTFE stopcock assembly
These economical PYREX® separatory funnels are designed to reduce replacement costs by providing replaceable components. The replaceable Polytetrafluorethylene (PTFE) Product Standard stopcock assembly has a simple screw thread locking nut and collar which ensure that the stopcock cannot fall out in use. The stem tip is also replaceable. Its ground end provides a better grip and helps prevent the tip from falling out.