60° Borosilicate Long Stem Funnels, Corning
Supplier: Corning
6140-75
470211-538CS 239.3 USD
470211-538
60° Borosilicate Long Stem Funnels, Corning
Funnels Filtration Funnels Filter Funnels
Quality, heavy walled borosilicate funnels with reinforced edge. The inside angle is approximately 60° and the stem length is 150 mm.