PYREX® Buchner Funnel with Fritted Disc
Supplier: Corning
36060-60M
470211-468CS 604.35 USD
470211-468
PYREX® Buchner Funnel with Fritted Disc
Funnels Filtration Funnels Büchner Funnels
PYREX®, borosilicate glass, with sintered glass disc.
  • No filter papers required
  • Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
  • Very good temperature resistance
Especially useful where filter paper would be attacked by material being filtered. Top edge is beaded.
Caution: Disc should not be subjected to pressure differential in excess of 1.01 bar (1 atm).