PYREX® Petri Culture Dishes
Supplier: Corning
3160-60
470211-460CS 1039.85 USD
470211-460
PYREX® Petri Culture Dishes
Culture Dishes Petri Dishes
Pyrex® dishes have beaded edges for strength. The tops are marked with blue enamel, and the bottoms with white enamel for easy sorting. Bottoms also have an arrow reference point for serial dilutions or orientation. Dish is not airtight. Autoclavable (wet or dry). Complete Dish Assembly