PYREX® Liebig Condenser
Supplier: Corning
2340-200
470211-398EA 172.85 USD
470211-398
PYREX® Liebig Condenser
Condensers
A fine borosilicate condenser with the inner tube sealed in.
The large-capacity water jacket ensures efficient cooling. The adapter end will accept a No. 3 rubber stopper.