Bottletop Vacuum/Filter Unit
Supplier: Corning
430769
470211-292CS 327 USD
470211-292
Bottletop Vacuum/Filter Unit
Filters Bottle Top Filters
This convenient system features a 90 mm Ø opening to filter faster through 0.22 μm membranes where autoclaving is not required.
Simply connect the unit to a vacuum pump or an aspirator filter pump. The base unscrews for use with bottles other than the one included. Capacity: 500 ml.