PYREX® Low-Form Beakers with Pour-Outs
Supplier: Corning
1010
470211-266CS 4134.35 USD
470211-266
PYREX® Low-Form Beakers with Pour-Outs
Beakers
Made of PYREX® borosilicate glass.
  • Excellent chemical resistance
  • Good chemical resistance
A sturdy, molded glass handle is secured to this 3 L beaker by a stainless steel strap which fits into a recess in the beaker wall. Hot liquids can be handled easily and safely.