Advanced Digital Free Fall Timer and Stopwatch
Supplier: OSAW INDUSTRIAL PRODUCTS SE
SV689
470211-168CS 1219.8 USD
470211-168
Advanced Digital Free Fall Timer and Stopwatch
Educational Materials Physics Educational Materials Mechanics Learning Activities
This advanced digital free fall timer and stopwatch uses modern electronics, piezo landing pad, and a push button launcher to accurately and conveniently perform free-fall experiments with fall distance of 50 cm to over 2 m.
Features include a large LED display, data storage up to five entries, a landing pad, push button, single on/off switch. Memory status, and sensor pulse and ball status shown by LEDs and built-in stopwatch.