Multi-Range DC Power Supply
Supplier: B & K Precision
9115
470210-672EA 3552.29 USD
470210-672
Multi-Range DC Power Supply
Power Supplies
This unit can replace multiple supplies on your lab bench.
  • Output up to 1200 W within 80 V/ 60A rating
  • Includes compact, high density, 1U rack mount form factor
  • High programming and read back resolution of 1 mV/1 mA
Unlike conventional power supplies with fixed output ratings, it automatically recalculates voltage/current limits for each setting. It provides 1200W output power in any Volt/Amp combination within the rated voltage (80V) and current (60A) limits. By providing greatly expanded choices of maximum power Volt/Amp combinations, users can cut down on the number of power supplies required and free up valuable bench space.