Disposable Bacti-Spreaders
Supplier: Excel Scientific
SPR-L-S10
470206-444BX 145.5 USD
470206-444
Disposable Bacti-Spreaders
Spreaders Cell Spreaders
These Bacti-Spreaders are ideal for beginning microbiology students.
  • Economical
  • Plastic
  • L Shape Simplifies Bacteria Distribution
  • Ideal for New Microbiology Students
They're an economical alternative to traditional metal Bacti-Spreaders.
The plastic instruments' L shape makes it easier to distribute bacteria over the entire plate.
Packaging: One package has 10 Bacti-Spreaders.