Microscope Slide Staining Dish with Rack
Supplier: DWK Life Sciences (KIMBLE)
900200
470205-616CS 779.15 USD
470205-616
Microscope Slide Staining Dish with Rack
Jars Staining Jars
Dries Slides Efficiently with Room for Multiple Slides
  • Quality slide making supplies
  • Slide staining tools
This quick-draining glass rack holds ten single slides, 19 slides arranged in a zigzag pattern, or 20 slides back to back. It comes with a glass cover.