Heavy-Duty Alcohol Burner
Supplier: DWK Life Sciences (KIMBLE)
237070
470205-612CS 379.65 USD
470205-612
Heavy-Duty Alcohol Burner
Heaters Spirit Lamps
This borosilicate glass burner features an extra-heavy flat base, ground-glass cover that fits snugly, and has no metal-to-metal contacts that can corrode.
  • Capacity: 100 ml
  • Size: 4"H
The cork stopper with metal top provides a better seal than metal screw tops.