Wall Mounted Eyewash Station
Supplier: WaterSaver Faucet
EW814
470205-492EA 665.55 USD
470205-492
Wall Mounted Eyewash Station
First Aid Supplies Eyewash Stations and Eyewash Bottles
This wall-mounted unit quickly and gently rinses contaminates away from both eyes simultaneously.
  • Stainless steel bowl for safe, simple use
  • Two heads deliver flood of water for hands-free eye rinsing
  • Heavy-duty pipe and fittings resist corrosion
Featuring two GS-Plus™ (Gentle Spray) Heads, this low-density polyethylene filter removes scale, deposits, and debris from the water and keeps water line impurities out of users' eyes.
A push handle activates the eye wash, which allows for hands free operation.
Certifications: ANSI-complaint identification sign included.