VWR® Bottle Tote™ Safety Carrier
Supplier: VWR International
470204-578
470204-578EA 152.2 USD
470204-578
VWR® Bottle Tote™ Safety Carrier
Carriers Bottle Carriers/Containers
Designed for safe transport of acids, alkalies, and solvents, a centering ring in the base assures 1" clearance between bottle and wall of the carrier.
  • Can be steam sterilized
  • Centering ring assures proper clearance for safe transport
Molded-in handle provides comfortable grip and will withstand 80lb. pull. Black carrier can be steam sterilized and accommodates 2.5 L (5-pint) or 4L (1-gallon) bottles.