VWR® One Block Heater

Supplier: VWR International
470204-396
470204-396EA 90.85 USD
470204-396
VWR® One Block Heater
Block Heaters Blocks
One Block Heaters are ideal for a variety of molecular biology procedures such as enzyme reactions, enzyme deactivations, and culture incubation.

  • Temperature Range: Ambient +5 To 150°C
  • VAC: 120
  • Average Weight: 6 1/10 lb.
  • Length: 12½"
  • Width: 8"
  • Height: 3½"

The versatile heaters weigh 6 1/10 lb.

The temperature is fully adjustable, and the counter displays the heating time.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR