Benchtop Water Quality Meter, Sper Scientific

Supplier: Sper Scientific
860033
470204-194EA 743.58 USD
470204-194
Benchtop Water Quality Meter, Sper Scientific
Electrochemistry Meters pH and Multi-Parameter Meters
Reads pH, ORP (oxygen reduction potential), mV, conductivity, TDS (total dissolved solids), salinity and temperature.

  • Durable Housing
  • Easy to Read
  • Automatic or manual temperature compensation
  • Automatic or manual ranging, automatic buffer recognition
  • Digital and analog outputs
  • Min/max and 99 data points, data hold
  • 5 calibration points
  • Electrode status indication

Operates on AC voltage (no batteries). Also accepts any pH or ORP probe with a standard BNC connector.

Rugged housing with large LCD displaying the parameter being measured together with time, date and temperature (in °C or °F). Measures: ph, ORP (oxygen reduction potential - SOLD SEPARATELY), mV, Conductivity, TDS (total dissolved solids), Salinity and Temperature.

pH Range: 0 - 14. Resolution: 0.01pH. Accuracy: ±0.02pH.
Conductivity Range: 0 - 19.9, 199.9, 1999 μS/cm; 0 - 19.9, 199.9 mS/cm. Resolution: 0.01, 0.1, 1μS/cm; 0.01, 0.1mS/cm. Accuracy: ±1% fs + 1d
TDS Range: 0 - 19.9, 199.9, 1999 ppm; 0 - 19.9, 199. 9 ppt. Resolution: 0.01, 0.1, 1ppm; 0.01, 0.1ppt. Accuracy: ±1% fs + 1d
Salinity Range: 0 - 11.38, 80 ppt. Resolution: 0.01, 0.1ppt. Accuracy: ±1% fs + 1d.
Temperature Range: -5°C - 80C; 23 - 176°F. Resolution: 0.1°F/°C. Accuracy: ±0.9°F/0.5°C

Ordering information: 5-year warranty. Comes ready to use with ATC conductivity and pH electrodes, multiple electrodes holder arm, AC adaptor, computer cable, and software.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR