Advanced pH Meter Kit, Sper Scientific
Supplier: Sper Scientific
850056
470204-174EA 445 USD
470204-174
Advanced pH Meter Kit, Sper Scientific
Electrochemistry Meters pH and Multi-Parameter Meters
Reads either pH (in 0.01 resolution) or mV in large digits, with time, date, and temperature (in °C or °F) shown simultaneously on the lower display.
  • Measures multiple conditions
  • Durable Housing
  • Easy to Read
Also indicates stable reading, low battery and displays calibration data. Scroll 99 data points with time stamps and min/max readings directly on the backlit LCD. Features hold, up to 5 point calibration, automatic buffer recognition, manual or automatic temperature compensation, and a USB port.
pH Range: 0 - 14. Resolution: 0.01pH. Accuracy: ±0.02pH.
mV Range: -499 to +499mV; -199.9 to +199.9mV. Resolution: 1mV; 0.1mV. Accuracy: ±2mV; ±0.2mV
Temperature Range: 23 - 176°F; -5 - 80°C Resolution: 0.1°F/°C. Accuracy: ±2°F; ±1°C
Ordering information: 5-year warranty. Kit includes Advanced pH Meter model 850055 in a hard carrying case complete with a detachable temperature compensated pH probe with 3½’ cable, pH buffers 4,7,10 and distilled water.