Conductivity Pen, Large Display, Sper Scientific
Supplier: Sper Scientific
850037
470204-162EA 118.3 USD
470204-162
Conductivity Pen, Large Display, Sper Scientific
Electrochemistry Meters Conductivity Meters
Highly stable and accurate readings with large LCD display. Designed for simple one-hand operation.
  • Designed for simple one-hand operation
  • Simultaneously displays the parameter being measured and temperature in °C or °F
  • Features automatic temperature compensation, 2-pt. calibration, auto power off, hold function, and a low battery indicator
  • Case is IP65 waterproof, and will float if accidentally dropped into the water
Conductivity Range: 0 - 1999μS; 0 - 19.99mS. Resolution: 1μS; 0.01mS. Accuracy: ±1% fs + 1d.
Temperature Range: 32 - 122 °F/ 0 - 50 °C. Resolution: 0.1 °F /°C. Accuracy: ±2 °F /1°C
Ordering information: 1-year warranty. Comes with 4 button-cell batteries and a built-in probe with protective cap.