Salinity/Temperature Pen, Large Display, Sper Scientific
Supplier: Sper Scientific
850036
470204-160EA 90.63 USD
470204-160
Salinity/Temperature Pen, Large Display, Sper Scientific
Electrochemistry Meters Salinity Meters
Simultaneously displays the parameter being measured and temperature in °C or °F.
  • Highly stable and accurate readings with large LCD display
  • Designed for simple one-hand operation
  • Automatic temperature compensation, 2-pt. calibration
  • Auto power off, hold function, low battery indicator
  • Case is IP65 waterproof, and will float if accidentally dropped into the water
Salinity Range: 0 - 10ppt; 10 - 70ppt. Resolution: 0.01ppt; 0.1ppt. Accuracy: ±1% fs + 1d.
Temperature Range: 32 - 122°F/0 - 50°C. Resolution: 0.1°F/°C. Accuracy: ±2°F/1°C
Ordering information: 1-year warranty. Comes with 4 button-cell batteries and a built-in probe with protective cap.