Infrared Thermometer with LED Light Pen
Supplier: Sper Scientific
800100
470204-106EA 85.68 USD
470204-106
Infrared Thermometer with LED Light Pen
Thermometers Digital Thermometers
Collect Temperature Values with the Press of a Button.
  • Provides 35 hours of continuous use
  • Thermometer is combined with a bright LED
  • Ideal for field use
With a response time of one second, this non–contact infrared thermometer quickly and accurately scans for temperature in Celsius and Fahrenheit. The thermometer end is protected by a cap when not in use, while the other side of the pen features a super bright white LED. Especially useful in the field, this versatile tool can provide 35 hours of continuous temperature measurement. Includes one AAAMore Product Information