Mason’s Hygrometer
Supplier: Sper Scientific
736740
470204-090EA 27.44 USD
470204-090
Mason’s Hygrometer
Hygrometers Analog Hygrometers
Easily measure humidity
  • Instructions included
  • Safe, does not contain mercury
One thermometer bulb is kept constantly wet by a siphon reservoir while the other gives a dry temperature reading. Humidity is obtained by comparing the two readings on the enclosed chart. The red liquid masons hygrometer uses a safe and easy to read red liquid in place of the usual mercury. Dual scales of -10/50°C and 10/120°F.