Salt Refractometer
Supplier: Sper Scientific
300011
470204-080EA 180.55 USD
470204-080
Salt Refractometer
Refractometers Benchtop Refractometers
The salt refractometer reduces the amount of samples needed.
  • No Manual Corrections Needed
  • Hard Carrying Case Included
  • Useful for Food Science
  • Salinity Range: 0-100 ppt; 1.000-1.070 SG
  • Resolution: 1 ppt; 0.001 SG
  • Accuracy: ±0.2%
The Refractometer features Automatic Temperature Compensation (ATC), which makes manual temperature corrections unnecessary. It measures salt content in sea water, aquariums, breeding ponds, or pickling-brines. While the scale is calibrated for sodium chloride, it can also be used for calcium chloride, barium cobalt, magnesium, cesium, potassium, sodium chloride, cupric sulfate, potassium iodide, citric acid, and acetic acid (vinegar) when tested against a known standard.
To operate, place a drop or two of sample on the prism, and read the results.
Ordering information: The Refractometer is supplied with a hard carrying case, calibration tool, and five-year manufacturer's warranty.