PCR Tubes
Supplier: Sorenson Bioscience
16950
470204-046CS 877 USD
470204-046
PCR Tubes
Tubes PCR Tubes
PCR tubes with attached caps.
  • Natural polypropylene
  • Optically clear cap
  • No graduation
Non-sterile pack.
Certified free from RNase/DNase, PCR inhibitor, human DNA and pyrogen.
Packaging: 1000 tubes/bag, 10 bags/case.