Inoculating Needle with Nickel-Chromium Wire and Aluminum Handle
Supplier: Science Center
3000
470203-984CS 114.5 USD
470203-984
Inoculating Needle with Nickel-Chromium Wire and Aluminum Handle
Spreaders Inoculating Loops and Needles
A 3" length of 24 gauge Ni/Cr wire set in a 6" L solid aluminum handle.