Promega PCR Nucleotide Mix
Supplier: PROMEGA SUNNYVALE INC SE
C1141
470203-864EA 142.6 USD
470203-864
Promega PCR Nucleotide Mix
Nucleic Acid Reagents Nucleotides
10 mM concentration each of dATP, dCTP, dGTP, and dTTP, for a total nucleotide concentration of 40 mM.
Note: Can be stored at 4°C for up to six weeks, or -20°C for longer.